missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHST15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597171
This item is not returnable.
View return policy
Description
Immunogen sequence: LQVGLYAVYL LDWLSVFDKQ QFLILRLEDH ASNVKYTMHK VFQFLNLGPL SEKQEALMTK SPASNARRPE DRNLGPMWPI TQKILRDFYR PFNARLAQVL ADEAFAWKTT; Positive Samples: Raji, Rat heart; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.
GALNAC4S-6ST is a sulfotransferase that transfers sulfate from 3'-phosphoadenosine 5'-phosphosulfate (PAPS) to the C-6 hydroxyl group of the GalNAc 4-sulfate residue of chondroitin sulfate A and forms chondroitin sulfate E containing GlcA-GalNAc(4, 6-SO(4)) repeating units. It also transfers sulfate to a unique non-reducing terminal sequence, GalNAc(4SO4)-GlcA(2SO4)-GalNAc(6SO4), to yield a highly sulfated structure similar to the structure found in thrombomodulin chondroitin sulfate. GALNAC4S-6ST may also act as a B-cell receptor involved in BCR ligation-mediated early activation that mediate regulatory signals key to B-cell development and/or regulation of B-cell-specific RAG expression; however such results are unclear in vivo.
Specifications
| CHST15 | |
| Polyclonal | |
| Unconjugated | |
| CHST15 | |
| 4631426J05Rik; B cell RAG associated protein; B cell RAG associated protein (GALNAC4S-6ST); B-cell RAG-associated gene protein; Brag; carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15; carbohydrate sulfotransferase 15; Chst15; DKFZp781H1369; GalNAc 4-sulfate 6-O-sulfotransferase; GALNAC4S6ST; GALNAC4S-6ST; GalNAcS-6ST; hBRAG; KIAA0598; MAd5; MGC34346; mKIAA0598; N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase; RP11-47G11.1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 286974, 51363 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.77 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q7LFX5, Q8CHI9 | |
| CHST15 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 452-561 of human CHST15 (NP_0569762). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction