missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHST14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567187
This item is not returnable.
View return policy
Description
Immunogen sequence: GILAEMKPLPL HPPGREGTAW RGKAPKPGGL SLRAGDADLQ VRQDVRNRTL RAVCGQPGMP RDPWDLPV Highest antigen sequence identity to the following orthologs - mouse 81%, rat 78%.
Catalyzes the transfer of sulfate to position 4 of the N-acetylgalactosamine (GalNAc) residue of dermatan sulfate. Plays a pivotal role in the formation of 4-0-sulfated IdoA blocks in dermatan sulfate. Transfers sulfate to the C-4 hydroxyl of beta1,4-linked GalNAc that is substituted with an alpha-linked iduronic acid (IdoUA) at the C-3 hydroxyl. Transfers sulfate more efficiently to GalNAc residues in -IdoUA-GalNAc-IdoUA- than in -GlcUA-GalNAc-GlcUA-sequences. Has preference for partially desulfated dermatan sulfate. Addition of sulfate to GalNAc may occur immediately after epimerization of GlcUA to IdoUA. GlcUA to IdoUA. Appears to have an important role in the formation of the cerbellar neural network during postnatal brain development.
Specifications
| CHST14 | |
| Polyclonal | |
| Unconjugated | |
| CHST14 | |
| 2600016L03Rik; ATCS; carbohydrate (dermatan 4) sulfotransferase 14; carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 14; carbohydrate sulfotransferase 14; CHST14; D4ST1; D4ST-1; dermatan 4 sulfotransferase 1; dermatan 4-sulfotransferase 1; dermatan-4-sulfotransferase-1; EDSMC1; hD4ST1; HNK1ST; UNQ1925/PRO4400 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 113189 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NCH0 | |
| CHST14 | |
| Recombinant Human CHST14. Recombinant protein control fragment (Product #RP-105539). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction