missing translation for 'onlineSavingsMsg'
Learn More

CHST14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£222.00 - £513.00

Specifications

Antigen CHST14
Dilution Western Blot 1:250 - 1:500, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18202313
View Documents
Novus Biologicals
NBP2-57389
100 μL
£513.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18609476
View Documents
Novus Biologicals
NBP2-57389-25ul
25 μL
£222.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CHST14 Polyclonal specifically detects CHST14 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CHST14
Polyclonal
Rabbit
Lipid and Metabolism
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
113189
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GILAEMKPLPLHPPGREGTAWRGKAPKPGGLSLRAGDADLQVRQDVRNRTLRAVCGQPGMPRDPWDLPV
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot 1:250 - 1:500, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Unconjugated
RUO
Human
ATCS, carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 14, D4ST-1, D4ST1carbohydrate sulfotransferase 14, dermatan 4 sulfotransferase 1, Dermatan 4-sulfotransferase 1, EC 2.8.2.-, HD4ST, hD4ST1, HNK1ST
CHST14
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.