missing translation for 'onlineSavingsMsg'
Learn More

CHST14 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-47607

Product Code. 30557709

  • £245.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CHST14 Polyclonal antibody specifically detects CHST14 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CHST14
Polyclonal
Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:20-1:200
ATCS, carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 14, D4ST-1, D4ST1carbohydrate sulfotransferase 14, dermatan 4 sulfotransferase 1, Dermatan 4-sulfotransferase 1, EC 2.8.2.-, HD4ST, hD4ST1, HNK1ST
carbohydrate(N-acetylgalactosamine 4-0) sulfotransferase 14. (Uniprot# Q8NCH0)
100 μg
Lipid and Metabolism
113189
Store at -20°C. Avoid freeze-thaw cycles.
IgG
ELISA, Immunohistochemistry, Western Blot
Unconjugated
PBS, 50% Glycerol, pH (7.3)
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.