Learn More
CHST12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15341-1-AP-20UL
 View more versions of this product
Product Code. 16814484
  • £135.00 / 20µL
Description
Catalyzes the transfer of sulfate to position 4 of the N-acetylgalactosamine (GalNAc) residue of chondroitin and desulfated dermatan sulfate. Chondroitin sulfate constitutes the predomit proteoglycan present in cartilage and is distributed on the surfaces of many cells and extracellular matrices. Activity toward partially desulfated dermatan sulfate is however lower. Does not form 4, 6-di-O-sulfated GalNAc when chondroitin sulfate C is used as an acceptor.
Specifications
Specifications
CHST12
Polyclonal
Unconjugated
CHST12
C4S 2, C4ST 2, C4ST2, CHST12, Sulfotransferase Hlo
Rabbit
Antigen Affinity Chromatography
RUO
304322, 55501, 59031
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99LL3, Q9NRB3
Chst12
CHST12 Fusion Protein Ag7544
20 μL
Primary
Human, Rat, Mouse