missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHST12 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00055501-B01P
This item is not returnable.
View return policy
Description
CHST12 Polyclonal antibody specifically detects CHST12 in Human samples. It is validated for Western Blot
Specifications
| CHST12 | |
| Polyclonal | |
| Western Blot 1:500 | |
| NP_061111.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| C4S-2, C4ST2C4ST-2, carbohydrate (chondroitin 4) sulfotransferase 12, carbohydrate sulfotransferase 12, Chondroitin 4-O-sulfotransferase 2, Chondroitin 4-sulfotransferase 2, EC 2.8.2.5, Sulfotransferase Hlo | |
| CHST12 (NP_061111.1, 1 a.a. - 414 a.a.) full-length human protein. MTKARLFRLWLVLGSVFMILLIIVYWDSAGAAHFYLHTSFSRPHTGPPLPTPGPDRDRELTADSDVDEFLDKFLSAGVKQSDLPRKETEQPPAPGSMEESVRGYDWSPRDARRSPDQGRQQAERRSVLRGFCANSSLAFPTKERAFDDIPNSELSHLIVDDRHGAIYCYVPKVACTNWKRVMIVLSGSLLHRGAPYRDPLRIPREHVHNASAHLTFNKFWRRYGKLSRHLMKVKLKKYTKFLFVRDPFVRLISAFRSKFELENEEFYRKFAVPMLRLYANHTSLPASAREAFRAGLKVSFANFIQYLLDPHTEKLAPFNEHWRQVYRLCHPCQIDYDFVGKLETLDEDAAQLLQLLQVDRQLRFPPSYRNRTASSWEEDWFAKIPLAWRQQLYKLYEADFVLFGYPKPENLLRD | |
| 0.05 mg | |
| Cancer, Cell Biology, Cytoskeleton Markers, Endocrinology, Signal Transduction | |
| 55501 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction