missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CHST11 Antibody (4F1), Novus Biologicals™

Product Code. 18344099 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344099 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344099 Supplier Novus Biologicals™ Supplier No. H00050515M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CHST11 Monoclonal antibody specifically detects CHST11 in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHST11
Applications ELISA, Western Blot, Sandwich ELISA
Classification Monoclonal
Clone 4F1
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_060883
Gene Alias C4S-1, C4ST, C4St-1, C4ST1EC 2.8.2.5, carbohydrate (chondroitin 4) sulfotransferase 11, carbohydrate sulfotransferase 11, Chondroitin 4-O-sulfotransferase 1, Chondroitin 4-sulfotransferase 1, DKFZp667A035, FLJ41682, HSA269537, IgH/CHST11 fusion
Host Species Mouse
Immunogen CHST11 (NP_060883, 230 a.a. ∽ 337 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RKGDDVKFEEFVAYLIDPHTQREEPFNEHWQTVYSLCHPCHIHYDLVGKYETLEEDSNYVLQLAGVGSYLKFPTYAKSTRTTDEMTTEFFQNISSEHQTQLYEVYKLD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50515
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.