missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHST1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542740
This item is not returnable.
View return policy
Description
Peptide sequence: SFVGQLFNQH LDVFYLFEPL YHVQNTLIPR FTQGKSPADR RVMLGASRDL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
Carbohydrate sulfotransferase 1 (CHST1), also referred to as KSGal6ST or KSST, is a keratan sulfate sulfotransferase. It is predominantly expressed in brain and skeletal muscle and localizes to the trans-Golgi network. CHST1 is responsible for mediating the sulfation of keratan in the cornea, which is important in maintaining corneal transparency. In particular, CHST1 catalyzes the transfer of sulfate groups from 3'-phosphoadenosine 5'-phosphosulfate to position six of internal or terminal galactose residues (preferentially the residues adjacent to sulfated GlcNAc) on growing keratan sulfate chains. CHST1 also contributes to the generation of L-selectin ligands. Mutations in the CHST1 gene may play a role in macular corneal dystrophy (MCD).
Specifications
| CHST1 | |
| Polyclonal | |
| Unconjugated | |
| CHST1 | |
| 2610008E20Rik; AW125896; C6ST; carbohydrate (chondroitin 6/keratan) sulfotransferase 1; carbohydrate (keratan sulfate Gal-6) sulfotransferase 1; carbohydrate sulfotransferase 1; chst1; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 1; Gst1; GST-1; hypothetical protein LOC445124; keratan sulfate Gal-6 sulfotransferase; Keratan sulfotransferase; KS6ST; KSGal6ST; KSST; N-acetylglucosamine-6-O-sulfotransferase 1; sulfo1; sulfotransferase1; zgc:100904; zgc:100904 protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8534 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O43916 | |
| CHST1 | |
| Synthetic peptide directed towards the N-terminal of human CHST1 (aa 74-123). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction