missing translation for 'onlineSavingsMsg'
Learn More

CHS-828, MedChemExpress

Product Code. p-200098796
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16495897

Brand: Medchem Express HY10079

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CHS-828 (GMX1778) is a competitive inhibitor of nicotinamide phosphoribosyltransferase (NAMPT), with an IC50 less than 25 nM. CHS-828 (GMX1778) exerts a cytotoxic effect by decreasing the cellular level of NAD+ and exhibits a potent anticancer activity.

CHS-828 (GMX1778) is a competitive inhibitor of nicotinamide phosphoribosyltransferase (NAMPT), with an IC50 less than 25 nM. CHS-828 (GMX1778) exerts a cytotoxic effect by decreasing the cellular level of NAD+ and exhibits a potent anticancer activity.

Specifications

Chemical Name or Material CHS-828
CAS 200484-11-3
Color White
Purity Grade Notes Research
Molecular Formula C19H22ClN5O
Quantity 5 mg
Synonym GMX1778
Solubility Information DMSO : ≥ 39 mg/mL (104.88 mM)
SMILES N#CN/C(NCCCCCCOC1=CC=C(C=C1)Cl)=N\C2=CC=NC=C2
Molecular Weight (g/mol) 371.86
Formula Weight 371.86
Percent Purity 99.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.