missing translation for 'onlineSavingsMsg'
Learn More

Chrysophenine (Technical Grade), TRC

Product Code. 30357962
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357962 250 mg -
30351186 500 mg 500mg
30366424 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357962 Supplier Toronto Research Chemicals Supplier No. TRCC433123250MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2870-32-8
Molecular Formula C30 H26 N4 O8 S2 . 2 Na
Molecular Weight (g/mol) 680.66
Synonym Benzenesulfonic acid, 2,2'-(1,2-ethenediyl)bis[5-[(4-ethoxyphenyl)azo]-, disodium salt (9CI), C.I. Direct Yellow 12 (6CI,7CI,8CI), Airedale Yellow CHD, Amanil Chrysophenine G, Atlantic Chrysophenine YA, Aurophenine O, Benzanil Yellow CH, Benzo Yellow LG, C.I. 24895, Calcomine Brilliant Yellow, Chrome Leather Yellow CH, Chrysophenine, Chrysophenine ESP, Chrysophenine Extra, Chrysophenine G, Chrysophenine G Extra, Chrysophenine GP, Chrysophenine J, Chrysophenine KG Conc, Chrysophenine NS, Chrysophenine Y, Chrysophenine YA-CF, Chrysophenine Yellow, Chrysophrenin G, Cotton Yellow CH, Diazamine Yellow C, Diphenyl Chrysoine 3G, Diphenyl Chrysoine 3GP, Direct Chrysophenine G, Direct Frosted Yellow G, Direct Frozen Yellow G, Direct Yellow 12, Direct Yellow C, Direct Yellow CHD, Direct Yellow CV, Direct Yellow G, Erie Yellow Y, Fenamin Yellow CP, Fixanol Yellow 3G, Kayafect Yellow S, Kayaku Chrysophenine, Kayaku Chrysophenine GE, Kayaku Chrysophenine GN, Kayaku Chrysophenine GX, Mitsui Chrysophenine G, Mitsui Chrysophenine G 400, Mitsui Chrysophenine KG, NSC 47745, Nylomine Acid Yellow C 4R, Peeramine Chrysophenine YA, Pontamine Yellow CH, Shikiso Chrysophenine GX, Tertrodirect Yellow CG, Vondacel Yellow CG
IUPAC Name disodium;5-[(E)-(4-ethoxyphenyl)diazenyl]-2-[(E)-2-[4-[(E)-(4-ethoxyphenyl)diazenyl]-2-sulfonatophenyl]ethenyl]benzenesulfonate
SMILES [Na+].[Na+].CCOc1ccc(cc1)N=Nc2ccc(\C=C\c3ccc(cc3S(=O)(=O)[O-])N=Nc4ccc(OCC)cc4)c(c2)S(=O)(=O)[O-]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C30H28N4O8S2.2Na/c1-3-41-27-15-11-23(12-16-27)31-33-25-9-7-21(29(19-25)43(35,36)37)5-6-22-8-10-26(20-30(22)44(38,39)40)34-32-24-13-17-28(18-14-24)42-4-2;;/h5-20H,3-4H2,1-2H3,(H,35,36,37)(H,38,39,40);;/q;2*+1/p-2/b6-5+,33-31?,34-32?;;
Formula Weight 680.0987
Chemical Name or Material Direct Yellow 12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.