missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chrysophanic Acid, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Chrysophanol |
| CAS | 481-74-3 |
| Molecular Formula | C15 H10 O4 |
| Quantity | 50 mg |
| Synonym | 9,10-Anthracenedione, 1,8-dihydroxy-3-methyl- (9CI, ACI), 1,8-Dihydroxy-3-methyl-9,10-anthracenedione (ACI), Anthraquinone, 1,8-dihydroxy-3-methyl- (8CI), 1,8-Dihydroxy-3-methyl-9,10-anthraquinone, 1,8-Dihydroxy-3-methylanthraquinone, 2-Methyl-4,5-dihydroxyanthraquinone, 3-Methyl-1,8-dihydroxyanthraquinone, 3-Methylchrysazin, 4,5-Dihydroxy-2-methylanthraquinone, C.I. 75400, C.I. Natural Yellow 23, Chrysophanic acid, Chrysophanol, NSC 37132, NSC 646567, Turkey Rhubarb |
| InChI Formula | InChI=1S/C15H10O4/c1-7-5-9-13(11(17)6-7)15(19)12-8(14(9)18)3-2-4-10(12)16/h2-6,16-17H,1H3 |
| SMILES | Cc1cc(O)c2C(=O)c3c(O)cccc3C(=O)c2c1 |
| IUPAC Name | 1,8-dihydroxy-3-methylanthracene-9,10-dione |
| Molecular Weight (g/mol) | 254.24 |
| Formula Weight | 254.0579 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction