missing translation for 'onlineSavingsMsg'
Learn More

Chrysophanic Acid, TRC

Product Code. 30359608
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30359608

Brand: Toronto Research Chemicals TRCC4326755MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chrysophanol
CAS 481-74-3
Molecular Formula C15 H10 O4
Quantity 5 mg
Synonym 9,10-Anthracenedione, 1,8-dihydroxy-3-methyl- (9CI, ACI), 1,8-Dihydroxy-3-methyl-9,10-anthracenedione (ACI), Anthraquinone, 1,8-dihydroxy-3-methyl- (8CI), 1,8-Dihydroxy-3-methyl-9,10-anthraquinone, 1,8-Dihydroxy-3-methylanthraquinone, 2-Methyl-4,5-dihydroxyanthraquinone, 3-Methyl-1,8-dihydroxyanthraquinone, 3-Methylchrysazin, 4,5-Dihydroxy-2-methylanthraquinone, C.I. 75400, C.I. Natural Yellow 23, Chrysophanic acid, Chrysophanol, NSC 37132, NSC 646567, Turkey Rhubarb
InChI Formula InChI=1S/C15H10O4/c1-7-5-9-13(11(17)6-7)15(19)12-8(14(9)18)3-2-4-10(12)16/h2-6,16-17H,1H3
SMILES Cc1cc(O)c2C(=O)c3c(O)cccc3C(=O)c2c1
IUPAC Name 1,8-dihydroxy-3-methylanthracene-9,10-dione
Molecular Weight (g/mol) 254.24
Formula Weight 254.0579
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.