missing translation for 'onlineSavingsMsg'
Learn More

Chrysin, 99.4%, For HPLC analysis, MP Biomedicals™

Product Code. 11423192 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423192 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11423192 Supplier MP Biomedicals Supplier No. 0215498505
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Naturally occurring flavonoid used for research or further manufacturing applications.

Specifications

CAS 480-40-0
Color Yellow
Assay Percent Range 99.4%
Molecular Formula C15H10O4
MDL Number MFCD00006593
Quantity 5 g
Synonym 4-nitrophenyl-beta-d-glucopyranoside, pnpg, 4-nitrophenyl beta-d-glucopyranoside, 1-o-p-nitrophenyl-d-glucose, 4-nitrophenyl beta-d-glucoside, p-nitrophenyl beta-d-glucopyranoside, p-nitrophenyl beta-d-glucoside, glucoside, p-nitrophenyl, p-nitrophenyl-beta-glucoside, p-nitrophenyl beta-glucoside
InChI Key RTIXKCRFFJGDFG-UHFFFAOYSA-N
SMILES OC1=CC(O)=C2C(=O)C=C(OC2=C1)C1=CC=CC=C1
IUPAC Name 5,7-dihydroxy-2-phenyl-4H-chromen-4-one
Molecular Weight (g/mol) 254.24
PubChem CID 92930
Formula Weight 254.24
Grade Research
Physical Form Powder
Chemical Name or Material Chrysin, For HPLC analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.