missing translation for 'onlineSavingsMsg'
Learn More

Chrysin-7-O-glucuronide, MedChemExpress

Product Code. 16471718
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471718 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471718 Supplier Medchem Express Supplier No. HYN2376

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Chrysin-7-O-glucuronide is a flavonoid extracted from Scutellaria baicalensis, with antioxidant activity.

Chrysin-7-O-glucuronide is a flavonoid extracted from Scutellaria baicalensis, with antioxidant activity.

Specifications

Chemical Name or Material Chrysin-7-O-glucuronide
CAS 35775-49-6
Color White
Purity Grade Notes Research
Molecular Formula C21H18O10
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (116.18 mM; Need ultrasonic)
SMILES O=C1C2=C(O)C=C(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(O)=O)C=C2OC(C4=CC=CC=C4)=C1
Molecular Weight (g/mol) 430.36
Formula Weight 430.36
Percent Purity 99.31%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.