missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chrysanthemic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Chrysanthemic acid |
| CAS | 10453-89-1 |
| Molecular Formula | C10 H16 O2 |
| Quantity | 25 mg |
| Synonym | Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propen-1-yl)-, Chrysanthemummonocarboxylic acid (6CI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)- (9CI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methylpropenyl)- (8CI), 2,2-Dimethyl-3-(2-methyl-1-propen-1-yl)cyclopropanecarboxylic acid, 2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanecarboxylic acid, 2,2-Dimethyl-3-(2-methyl-propenyl)-cyclopropanecarboxylic acid, 2,2-Dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropane-1-carboxylic acid, 3,3-Dimethyl-2-(2-methyl-1-propenyl)cyclopropanecarboxylic acid, 3-Isobutenyl-2,2-dimethylcyclopropanecarboxylic acid, Chrysanthemic acid, Chrysanthemumic acid, NSC 11779 |
| InChI Formula | InChI=1S/C10H16O2/c1-6(2)5-7-8(9(11)12)10(7,3)4/h5,7-8H,1-4H3,(H,11,12) |
| SMILES | CC(=CC1C(C(=O)O)C1(C)C)C |
| IUPAC Name | 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylic acid |
| Molecular Weight (g/mol) | 168.23 |
| Formula Weight | 168.115 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?