missing translation for 'onlineSavingsMsg'
Learn More

Chrysanthemic Acid, TRC

Product Code. 30350970
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350970 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350970 Supplier Toronto Research Chemicals Supplier No. TRCC903440100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chrysanthemic acid
CAS 10453-89-1
Molecular Formula C10 H16 O2
Quantity 100 mg
Synonym Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propen-1-yl)-, Chrysanthemummonocarboxylic acid (6CI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)- (9CI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methylpropenyl)- (8CI), 2,2-Dimethyl-3-(2-methyl-1-propen-1-yl)cyclopropanecarboxylic acid, 2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanecarboxylic acid, 2,2-Dimethyl-3-(2-methyl-propenyl)-cyclopropanecarboxylic acid, 2,2-Dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropane-1-carboxylic acid, 3,3-Dimethyl-2-(2-methyl-1-propenyl)cyclopropanecarboxylic acid, 3-Isobutenyl-2,2-dimethylcyclopropanecarboxylic acid, Chrysanthemic acid, Chrysanthemumic acid, NSC 11779
InChI Formula InChI=1S/C10H16O2/c1-6(2)5-7-8(9(11)12)10(7,3)4/h5,7-8H,1-4H3,(H,11,12)
SMILES CC(=CC1C(C(=O)O)C1(C)C)C
IUPAC Name 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylic acid
Molecular Weight (g/mol) 168.23
Formula Weight 168.115
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.