missing translation for 'onlineSavingsMsg'
Learn More

Chrysamine G Disodium Salt, TRC

Product Code. 30137793
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137793 25 mg 25mg
30133911 100 mg 100mg
30005271 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137793 Supplier Toronto Research Chemicals Supplier No. TRCC43302025MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6472-91-9
Molecular Formula C26H16N4Na2O6
Molecular Weight (g/mol) 526.41
Synonym C.I. Direct Yellow 1, C.I. Direct Yellow 1, Disodium Salt, Chrysamine, Brasilamina Yellow G, C.I. 22250, Chrysamine A, Chrysamine Yellow M, Diazine Yellow 2G, Direct Yellow CPS, Disodium diphenyl-4,4'-bis(azosalicylate), Kayaku Chrysamine G, 3,3'-[[1,1'-Biphenyl]-4,4'-diylbis(azo)]bis[6-hydroxy-benzoic Acid Disodium Salt
IUPAC Name sodium 5,5'-((1E,1'E)-[1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(2-hydroxybenzoate)
SMILES O=C(O[Na])C1=C(O)C=CC(/N=N/C2=CC=C(C3=CC=C(/N=N/C4=CC(C(O[Na])=O)=C(O)C=C4)C=C3)C=C2)=C1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C26H18N4O6.2Na/c31-23-11-9-19(13-21(23)25(33)34)29-27-17-5-1-15(2-6-17)16-3-7-18(8-4-16)28-30-20-10-12-24(32)22(14-20)26(35)36;;/h1-14,31-32H,(H,33,34)(H,35,36);;/q;2*+1/p-2/b29-27+,30-28+;;
Formula Weight 526.09
Chemical Name or Material Chrysamine G Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.