missing translation for 'onlineSavingsMsg'
Learn More

Chromyl chloride, 99.99% (metals basis)

Product Code. 11398498
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398498 10 g 10g
11318508 2 g 2g
11308508 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11398498 Supplier Thermo Scientific Alfa Aesar Supplier No. 040517.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14977-61-8 | Cl2CrH4O2 | 158.926 g/mol

Highly spin-polarized chromium dioxide (CrO2) thin films were deposited on 100 TiO2 substrates by chemical vapor deposition using chromyl chloride as a precursor. Chromyl chloride (Cr02C12) reacts with cyclohexane solvent at 75 OC to give a dark precipitate along with chlorocyclohexane and a small amount of cyclohexene (in 10.0 and ca. 0.3% yields based on chromium). Chromyl chloride, CrO2C12, oxidizes cyclooctane, isobutane, and toluene under mild conditions (25-60 C). The reactions give chlorinated products (chlorocyclooctane, tert- butyl chloride, and benzyl chloride) and a dark chromium-containing precipitate. Used for Organic oxidations and chlorination, chromium coordination complexes, catalyst for polymerization of olefins. Chromyl chloride in carbon tetrachloride solution reacts in the cold with cyclohexene, cyclopentene, and 1-hexene to give the various isomeric chlorohydrins as the major products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Organic oxidations and chlorination, chromium coordination complexes, catalyst for polymerization of olefins
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14977-61-8
Molecular Formula Cl2CrH4O2
Molecular Weight (g/mol) 158.926
MDL Number MFCD00010951
InChI Key HOCPFYCTFLRIAK-UHFFFAOYSA-L
Synonym chromyl chloride
PubChem CID 101943071
IUPAC Name chromium(2+);dichloride;dihydrate
SMILES O.O.[Cl-].[Cl-].[Cr+2]

Specifications

Boiling Point 117°C
Melting Point -96.5°C
Physical Form Liquid
Quantity 10 g
UN Number UN1758
Merck Index 14,2248
Solubility Information Soluble in carbon tetrachloride,carbon disulfide,benzene,nitrobenzene,chloroform,and POCL3. Insoluble in water(Reacts).
Formula Weight 154.9
Percent Purity 99.99%
Odor Pungent, makes eyes water
Sensitivity Moisture sensitive
Density 1.911g/mL
Assay (metals basis)
Chemical Name or Material Chromyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.