missing translation for 'onlineSavingsMsg'
Learn More

Chromplete™ Methanol, HPLC/GC, meets ACS/USP analytical requirements

Product Code. 15990711 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
Unit Size:
1 L
2.5 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15990711 1 L 1 L
15900721 2.5 L 2.5 L
15910721 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15990711 Supplier Fisher Scientific Chemical Div Supplier No. T001021000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Methanol is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry. | CAS: 67-56-1 | CH4O | 32.04 g/mol

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Methanol is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry.

Chromplete Methanol meets the analytical requirements of USP and is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • Pesticide Analysis by GC or HPLC
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Absorbance ≤1.00, ≤ 0.23, ≤ 0.11, ≤ 0.01
CAS Min % 0.999
Color ≤ 5 APHA
Density 0.775 – 0.780
Quantity 1 L
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation ≤ 1.0 ppm
Titratable Acid ≤0.0003 meq/g
Titratable Base ≤0.0002 meq/g
UN Number 1230
Water ≤0.07%
Boiling Point 148.5°F
Formula Weight 32.04
Percent Purity 99.9%
Grade Chromplete
Assay Minimum, GC-FID
Chemical Name or Material Methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chromplete Methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: methanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.