missing translation for 'onlineSavingsMsg'
Learn More

Chromotrope FB, Thermo Scientific Chemicals

Product Code. 11426036
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3567-69-9
Molecular Formula C20H12N2Na2O7S2
Molecular Weight (g/mol) 502.42
MDL Number MFCD00003978
InChI Key OJNIRNOPCQQNRY-JZFXEGLSSA-L
Synonym Acid Red 14; C.I. 14720
PubChem CID 44135716
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=C\C(=N/NC2=C3C=CC=CC3=C(C=C2)S([O-])(=O)=O)C(=O)C2=CC=CC=C12

Specifications

IUPAC Name disodium;4-oxo-3-[(4-sulfonatonaphthalen-1-yl)hydrazinylidene]naphthalene-1-sulfonate
Formula Weight 502.42
Quantity 10 g
Chemical Name or Material Chromotrope FB

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'