Learn More
Chromotrope 2R
Description
Employed as an indicator for complexometry. It is also applied as a stain used in modified Gomori Trichrome stain. Used as a stain for cytology.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Employed as an indicator for complexometry. It is also applied as a stain used in modified Gomori Trichrome stain. Used as a stain for cytology.
Solubility
Soluble in water (1 mg/ml).
Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry place.
Specifications
Specifications
| Molecular Formula | C16H10N2Na2O8S2 |
| Solubility Information | Soluble in water (1mg/ml). |
| Formula Weight | 468.37 |
| Quantity | 100 g |
| Chemical Name or Material | Chromotrope 2R |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.