missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chromotrope 2R, pure

Product Code. 10144210 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10144210 25 g Glass bottle 25g
10702305 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10144210 Supplier Thermo Scientific Chemicals Supplier No. 192010250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4197-07-3 | C16H10N2Na2O8S2 | 468.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4197-07-3
Molecular Formula C16H10N2Na2O8S2
Molecular Weight (g/mol) 468.36
MDL Number MFCD00003955
InChI Key XTJONEUTTZZRAB-VZRGYXGKSA-L
Synonym 4,5-Dihydroxy-3-phenylazo-2,7-naphthalenedisulfonic acid, disodium salt, C.I. 16570, 4, 5-Dihydroxy-3-phenylazo-2, 7-naphthalenedisulfonic acid
PubChem CID 44135388
IUPAC Name disodium;5-hydroxy-4-oxo-3-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=CC(=C3C2=O)O)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Beilstein 16, 301
Infrared Spectrum Authentic
Formula Weight 468.35
Grade Extra Pure
Packaging Glass bottle
Specific Extinction 360 min. (509nm, water)
Color Brown/Red
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Chromotrope 2R
compliance-icons
Product Identifier
  • Chromotrope 2R
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.