Learn More
Proteintech ChromoTek V5-Trap™ Magnetic Particles M-270 Kit
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Description
- V5-Trap Magnetic Particles M-270 Kit is used to immunoprecipitate V5-tagged fusion proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |