missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek V5-Trap™ Magnetic Particles M-270

Product Code. 17393533
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
This item is not returnable. View return policy

Product Code. 17393533

Brand: Proteintech v5td10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The ChromoTek V5-Trap™ Magnetic Particles M-270 consists of an anti-V5-tag Nanobody (VHH), which is covalently bound to Magnetic Particles M-270.

  • V5-Trap Magnetic Particles M-270 is used to immunoprecipitate V5-tagged fusion proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc.

Specifications

Content And Storage Upon receipt store at 4°C. Do not freeze.
For Use With (Application) IP, CoIP
Target V5-tag sequence GKPIPNPLLGLDST at the N-, C-terminal or internal tagging.
Label or Dye Magnetic Particles M-270
Species Alpaca
Sufficient For 10 Reactions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.