Learn More
Proteintech ChromoTek V5-Trap™ Magnetic Agarose Kit
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Description
- The ChromoTek V5-Trap™ Magnetic Agarose is an affinity resin for IP of V5-tagged proteins
- Consists of a anti V5 Nanobody/VHH coupled to magnetic agarose beads
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |