missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek V5-Trap™ Magnetic Agarose
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
Description
- Consists of a anti-V5 Nanobody/VHH coupled to magnetic agarose beads
Specifications
Specifications
| Content And Storage | Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. |
| For Use With (Application) | IP, CoIP, ChIP, RIP |
| Target | V5-tag sequence GKPIPNPLLGLDST at the N-, C-terminal or internal tagging. |
| Label or Dye | Magnetic Agarose |
| Species | Alpaca |
| Sufficient For | 400 Reactions |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction