missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek TurboGFP-Trap Magnetic Agarose

Product Code. 17343523
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
This item is not returnable. View return policy

Product Code. 17343523

Brand: Proteintech tbtma400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TurboGFP-Trap Magnetic Agarose is an affinity reagent for immunoprecipitation (IP) of TurboGFP fusion proteins.

  • Comprises an anti-TurboGFP VHH/ Nanobody coupled to magnetic agarose beads

Specifications

Content And Storage Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year.
For Use With (Application) IP, CoIP, ELISA
Target TurboGFP, CopGFP, Pontellina plumata, maxGFP, maxFP-Green
Label or Dye Magnetic Agarose
Species Alpaca
Sufficient For 400 Reactions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.