Learn More
Proteintech ChromoTek Spot-Trap™ Magnetic Particles M-270 Kit
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Description
- Spot-Trap™ Magnetic Particles M-270 are affinity beads for immunoprecipitation (IP) of Spot-tagged proteins
- Comprises an anti-Spot-Tag™ VHH/ Nanobody coupled to Magnetic Particles M-270
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |