Learn More
Proteintech ChromoTek Spot-Trap™ Magnetic Agarose Kit
Product Code. 17323323
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Product Code. 17323323
Brand: Proteintech etmak20
Kit including 20 reactions of Spot-Trap™ Magnetic Agarose and lysis, dilution, and wash buffers.
  • Spot-Trap™ Magnetic Agarose is an affinity reagent for immunoprecipitation (IP) of Spot-tagged proteins
  • Consists of an anti-Spot-Tag™ VHH/ Nanobody that is coupled to magnetic agarose beads
Specifications
Quantity 20 Reactions
Sufficient For 20 Reactions