missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek Spot-peptide

Product Code. 17313303
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 17313303

Brand: Proteintech ep1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Spot-peptide for elution of Spot-tagged fusion proteins bound to Spot-Cap.

  • Dilute Spot-peptide to 0.1 mM in PBS for elution of Spot-tagged
    proteins from Spot-Cap
  • Shipped at ambient temperature. Upon receipt store at -20°C
  • Aliquot upon reconstitution and store at -20°C
  • Avoid repeated freeze / thaw cycles
  • Reconstitution: Dissolve Spot-peptide in PBS (1 mg Spot-peptide in 140 μL PBS, 10 mg Spot-Peptide in 1.4 mL PBS) to a final concentration of 5 mM (7 mg/mL)

Specifications

Format Lyophilized Powder
Molecular Weight (g/mol) 1397 g/mol
Product Type Peptide of amino acid sequence PDRVRAVSHWSS
Quantity 1 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.