missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Spot-Label™ ATTO488
Spot VHH conjugated to ATTO 488.
£76.00 - £301.00
Specifications
Clonality | Monoclonal |
---|---|
For Use With (Application) | Immunofluorescence (IF), Western Blot |
Conjugate | ATTO488 |
Target Specificity | Spot-Tag (PDRVRAVSHWSS) |
Content And Storage | Shipped at ambient temperature. Upon receipt store at +4°C / 40°F.Stable for 6 months. Do not freeze. Protect from light. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17383283
|
Proteintech
EBA488-10 |
10 μL |
£76.00
10µL |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
17393283
|
Proteintech
EBA488-50 |
50 μL |
£301.00
50µL |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
Description
- anti-Spot-Tag VHH/ Nanobody conjugated to fluorescent dye for immunofluorescence, microscopy, and immunoblotting of Spot-tagged proteins
Specifications
Monoclonal | |
ATTO488 | |
Shipped at ambient temperature. Upon receipt store at +4°C / 40°F.Stable for 6 months. Do not freeze. Protect from light. |
Immunofluorescence (IF), Western Blot | |
Spot-Tag (PDRVRAVSHWSS) |