missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek Spot-Label™ Alexa Fluor™ 647

Product Code. 17363293
Click to view available options
Quantity:
10 μL
50 μL
Unit Size:
10µL
50µL
This item is not returnable. View return policy

Product Code. 17363293

Brand: Proteintech ebAF64710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Spot VHH conjugated to Alexa Fluor™ 647.

  • anti-Spot-Tag VHH/ Nanobody conjugated to fluorescent dye for immunofluorescence, microscopy, and immunoblotting of Spot-tagged proteins

Specifications

Clonality Monoclonal
Quantity 10 μL
For Use With (Application) Immunofluorescence (IF), Western Blot
Conjugate Alexa Fluor™ 647
Target Specificity Spot-Tag (PDRVRAVSHWSS)
Content And Storage Shipped at ambient temperature. Upon receipt store at +4°C / 40°F.Stable for 6 months. Do not freeze. Protect from light.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.