missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek Spin Columns

Product Code. 17313463
Click to view available options
Quantity:
10/Pk.
20/Pk.
50/Pk.
Unit Size:
Pack of 10
Pack of 20
Pack of 50
This item is not returnable. View return policy

Product Code. 17313463

Brand: Proteintech sct20

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Filtration units from PP and 20 μm PE frit for batch mode centrifugation based separation.

  • The spin columns allow rapid washing and clean elution of bound proteins.
    Spin columns simplify the pull-down of your protein with Nano-Traps coupled to agarose beads
  • The spin columns are designed for easy and convenient handling: For washing, you may discard the flow-through wash buffer; for elution, collect the flow-through after adding the elution buffer

Specifications

Quantity 20/Pk.
For Use With (Application) Rapid washing and clean elution of bound proteins
Type Spin Column
Sample Volume 500 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.