missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Spin Columns
Description
- The spin columns allow rapid washing and clean elution of bound proteins.
Spin columns simplify the pull-down of your protein with Nano-Traps coupled to agarose beads - The spin columns are designed for easy and convenient handling: For washing, you may discard the flow-through wash buffer; for elution, collect the flow-through after adding the elution buffer
Specifications
Specifications
| Quantity | 10/Pk. |
| For Use With (Application) | Rapid washing and clean elution of bound proteins |
| Type | Spin Column |
| Sample Volume | 500 μL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?