Learn More
Proteintech ChromoTek RFP-Trap™ Magnetic Agarose Kit
Product Code. 17393453
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Product Code. 17393453
Brand: Proteintech rtmak20
Kit including 20 reactions of RFP-Trap™ Magnetic Agarose and lysis, dilution, wash, and elution buffers.
  • The ChromoTek RFP-Trap™ Magnetic Agarose are affinity beads for IP of RFP-fusion proteins
  • Comprises a RFP Nanobody/ VHH coupled to magnetic agarose
Specifications
Quantity 20 Reactions
Sufficient For 20 Reactions