missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek RFP-Booster ATTO647N
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
Description
- anti-RFP VHH/ Nanobody conjugated to fluorophore for IF, of mCherry and other red fluorescent protein-fusion proteins
Specifications
Specifications
| Clonality | Monoclonal |
| Quantity | 10 μL |
| For Use With (Application) | Immunofluorescence (IF), Immunohistochemistry (IHC), Immunocytochemistry (ICC) |
| Conjugate | ATTO647N |
| Target Specificity | mCherry, mPlum, mRFP, mRFPruby, mKate2 |
| Content And Storage | Shipped at ambient temperature. Upon receipt store at +4°C. Stable for 6 month. Do not freeze. Protect from light. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction