missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek RFP-Booster Alexa Fluor™ 568
Click to view available options
Quantity:
10 μL
50 μL
Unit Size:
10µL
50µL
Description
- anti-RFP VHH/ Nanobody conjugated to fluorophore for IF, of mCherry and other red fluorescent protein-fusion proteins
Specifications
Specifications
| Clonality | Monoclonal |
| Quantity | 10 μL |
| For Use With (Application) | Immunofluorescence (IF), Immunohistochemistry (IHC), Immunocytochemistry (ICC) |
| Conjugate | Alexa Fluor™ 568 |
| Target Specificity | mCherry, mPlum, mRFP, mRFPruby, mKate2 |
| Content And Storage | Shipped at ambient temperature. Store at -20°C/-4°F. Avoid freeze-thaw cycles. Aliquot upon arrival. Protect from light. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction