Learn More
Proteintech ChromoTek pSpot3 Vector
Product Code. 17353323
Change view
Click to view available options
Quantity:
1.25 μg
Unit Size:
1.25µg
Product Code. Quantity Unit Size
17353323 1.25 μg 1.25µg
1 options
Product Code. 17353323 Supplier Proteintech Supplier No. ev3
  • ONLINE EXCLUSIVE PRICE
    £51.90
    £51.00 / 1.25µg
    Save £0.90
    1% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
pSpot3 vector, E.coli, Spot C-term, Amp., low expression.
  • The plasmid pSpot3 is an expression vector for Spot-Tag™ fusion proteins in E. coli. After cloning the protein of interest (POI) into the multiple cloning site (MCS) provided by pSpot3, the Spot-Tag™ (sequence: PDRVRAVSHWSS) will be fused to the C-terminus of the POI.
Specifications
Quantity 1.25 μg