Learn More
Proteintech ChromoTek PARP1-Chromobody™ Plasmid (TagGFP)
PARP1-Chromobody™ fused to TagGFP2.
Brand:  Proteintech XCG
Additional Details : Weight : 0.00240kg
Product Code. 17323563
  • £761.00 / 20µg
Estimated Shipment: 14-05-2024
to see stock.
Description
Description
  • DNA plasmid encoding for anti-PARP1 VHH (anti-PARP1 Nanobody) fused to TagGFP2
Specifications
Specifications
20 μg