missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek Nano-CaptureLigand™ Mouse IgG2a, Fc-specific VHH, Biotinylated

Product Code. 17393483
Change view
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17393483 10 μL 10µL
17303493 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17393483 Supplier Proteintech Supplier No. smsG2aB110
  • ONLINE EXCLUSIVE PRICE
    £103.81
    £102.00 / 10µL
    Save £1.81
    1% Off
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nano-CaptureLigand for mouse IgG2a: Fc-specific single domain antibody/VHH [CTK0108], biotinylated.

  • The Nano-CaptureLigand mouse IgG2a captures and immobilizes mouse IgG2a antibodies and Fc-fragments to avidin/streptavidin coated surfaces used in BLI, SPR, and ELISA assays
  • Consists of a biotinylated Nanobody that specifically binds to the mouse IgG2a subclass with high affinity and in a site-directed manner

Specifications

Quantity 10 μL
Target Specificity Fc-fragment of Mouse IgG2a
Conjugate Biotin
Clonality Monoclonal
Content And Storage Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F
For Use With (Application) BLI, SPR, ELISA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.