Learn More
Proteintech ChromoTek Myc-Trap™ Magnetic Agarose Kit
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Description
- The ChromoTek Myc-Trap™ Magnetic Agarose is an affinity resin for IP of Myc-fusion proteins
- Consists of a Myc Nanobody/VHH coupled to magnetic agarose beads
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |