missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Myc-Trap™ Magnetic Agarose
Description
- Consists of a Myc Nanobody/VHH coupled to magnetic agarose beads
Specifications
Specifications
| Content And Storage | Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. |
| For Use With (Application) | IP, CoIP, ChIP, RIP |
| Target | Myc-tag sequence motif EQKLISEEDL at the N-terminus, C-terminus, or internal site of the fusion protein. Endogenous c-myc is NOT bound. |
| Label or Dye | Magnetic Agarose |
| Species | Alpaca |
| Sufficient For | 20 Reactions |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?