missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek mNeonGreen-Trap Magnetic Agarose

Product Code. 17393393
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
This item is not returnable. View return policy

Product Code. 17393393

Brand: Proteintech ntma20

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The ChromoTek mNeonGreen-Trap Magnetic Agarose is an affinity resin for IP of mNeonGreen-fusion proteins.

  • Consists of a mNeonGreen Nanobody/ VHH coupled to magnetic agarose beads

Specifications

Content And Storage Upon receipt store at 4°C. Do not freeze.
For Use With (Application) IP, CoIP, ChIP, RIP
Target mNeonGreen, Branchiostoma lanceolatum
Label or Dye Magnetic Agarose
Species Alpaca
Sufficient For 20 Reactions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.