missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Halo-Trap Magnetic Agarose
Description
- Halo-Trap Magnetic Agarose is used to immunoprecipitate Halo-tagged fusion proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc. in the presence or absence of a covalently bound ligand
- The interaction between Halo-Trap and the Halo-tagged fusion protein is reversible
Specifications
Specifications
| Binding Capacity | 12.5 μg of recombinant Halo-tag per 25 μL bead slurry |
| Content And Storage | Upon receipt store at 4°C. Do not freeze. Stable for 1 year. Shipped at ambient temperature. |
| For Use With (Application) | IP, CoIP, ChIP, RIP |
| Target | Halo-tag (modified variant of the bacterial haloalkane dehalogenase enzyme from Rhodococcus rhodochrous) in the absence or presence of covalently bound chloralkane-based ligands |
| Label or Dye | Magnetic Agarose |
| Species | Alpaca |
| Sufficient For | 20 Reactions |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?