missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek Halo-Trap Magnetic Agarose

Product Code. 17303413
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
This item is not returnable. View return policy

Product Code. 17303413

Brand: Proteintech OTMA10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The ChromoTek Halo-Trap Magnetic Agarose consists of an anti-Halo-tag Nanobody (VHH), which is covalently bound to magnetic agarose beads.

  • Halo-Trap Magnetic Agarose is used to immunoprecipitate Halo-tagged fusion proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc. in the presence or absence of a covalently bound ligand
  • The interaction between Halo-Trap and the Halo-tagged fusion protein is reversible

Specifications

Binding Capacity 12.5 μg of recombinant Halo-tag per 25 μL bead slurry
Content And Storage Upon receipt store at 4°C. Do not freeze. Stable for 1 year. Shipped at ambient temperature.
For Use With (Application) IP, CoIP, ChIP, RIP
Target Halo-tag (modified variant of the bacterial haloalkane dehalogenase enzyme from Rhodococcus rhodochrous) in the absence or presence of covalently bound chloralkane-based ligands
Label or Dye Magnetic Agarose
Species Alpaca
Sufficient For 10 Reactions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.