missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek GFP-Trap™ Magnetic Particles M-270
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
Description
- GFP-Trap™ Magnetic Particles M-270 is highly recommended, when very large proteins/complexes are investigated, and magnetic separation is needed for IP
- Consists of a GFP VHH/ Nanobody coupled to Magnetic Particles M-270
Specifications
Specifications
| Binding Capacity | 10 μL slurry bind about 1 μg of recombinant GFP |
| Content And Storage | Upon receipt store at 4°C. Do not freeze. |
| For Use With (Application) | IP, CoIP, ChIP, RIP |
| Target | AcGFP, Clover, eGFP, Emerald, GFP, GFP5, GFP Envy, GFP S65T, mGFP, mPhluorin, PA-GFP, Superfolder GFP, TagGFP, TagGFP2, monomeric eGFP A206K, CFP, YFP, Citrine, eCitrine, eYFP, Venus, Ypet, BFP |
| Label or Dye | Magnetic Particles M-270 |
| Species | Alpaca |
| Sufficient For | 400 Reactions |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction