missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek GFP-Trap™ Magnetic Particles M-270

Product Code. 17353353
Change view
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For Unit Size
17353353 10 Reactions 10 reactions
17363353 100 Reactions 100 reactions
17373353 20 Reactions 20 reactions
17383353 200 Reactions 200 reactions
17393353 400 Reactions 400 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17353353 Supplier Proteintech Supplier No. GTD10
  • ONLINE EXCLUSIVE PRICE
    £262.58
    £248.00 / 10 reactions
    Save £14.58
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GFP-Trap™ Magnetic Particles M-270 for immunoprecipitation (IP) of GFP-tagged proteins.

  • GFP-Trap™ Magnetic Particles M-270 is highly recommended, when very large proteins/complexes are investigated, and magnetic separation is needed for IP
  • Consists of a GFP VHH/ Nanobody coupled to Magnetic Particles M-270

Specifications

Content And Storage Upon receipt store at 4°C. Do not freeze.
For Use With (Application) IP, CoIP, ChIP, RIP
Target AcGFP, Clover, eGFP, Emerald, GFP, GFP5, GFP Envy, GFP S65T, mGFP, mPhluorin, PA-GFP, Superfolder GFP, TagGFP, TagGFP2, monomeric eGFP A206K, CFP, YFP, Citrine, eCitrine, eYFP, Venus, Ypet, BFP
Label or Dye Magnetic Particles M-270
Species Alpaca
Sufficient For 10 Reactions
Binding Capacity 10 μL Slurry bind about 1 μg of Recombinant GFP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.