Learn More
Proteintech ChromoTek GFP-Trap™ Magnetic Agarose Kit
Product Code. 17363363
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Product Code. 17363363
Brand: Proteintech gtmak20
Kit including 20 reactions of GFP-Trap™ Magnetic Agarose and lysis, dilution, wash, and elution buffers.
  • The ChromoTek GFP-Trap™ Magnetic Agarose are affinity beads for IP of GFP-fusion proteins
  • Comprises a GFP Nanobody/ VHH coupled to magnetic agarose beads
Specifications
Quantity 20 Reactions
Sufficient For 20 Reactions