missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek GFP-Trap™ Magnetic Agarose
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
Description
- Comprises a GFP Nanobody/ VHH coupled to magnetic agarose beads
Specifications
Specifications
| Binding Capacity | 10 μL slurry bind more than 8 μg of recombinant GFP |
| Content And Storage | Upon receipt store at 4°C. Do not freeze. |
| For Use With (Application) | IP, CoIP, ChIP, RIP |
| Target | AcGFP, Clover, eGFP, Emerald, GFP, GFP5, GFP Envy, GFP S65T, mGFP, mPhluorin, PA-GFP, Superfolder GFP, TagGFP, TagGFP2, monomeric eGFP A206K, CFP, YFP, Citrine, eCitrine, eYFP, Venus, Ypet, BFP |
| Label or Dye | Magnetic Agarose |
| Species | Alpaca |
| Sufficient For | 400 Reactions |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering