missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek GFP-Booster ATTO594
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
Description
- anti-GFP VHH/ Nanobody conjugated to a fluorescent dye for IF/ microscopy
Specifications
Specifications
| Clonality | Monoclonal |
| Quantity | 10 μL |
| For Use With (Application) | Immunofluorescence (IF), Immunohistochemistry (IHC), Immunocytochemistry (ICC) |
| Conjugate | ATTO594 |
| Target Specificity | AcGFP, Citrine, CFP, eGFP, eYFP, GFP S65T, pHluorin, sfGFP, mClover (Clover A206K), TagGFP, tagGFP2, Venus, wtGFP, YFP |
| Content And Storage | Shipped at ambient temperature. Upon receipt store at +4™C. Stable for 6 month. Do not freeze. Protect from light. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction